Skip to Content
Merck
All Photos(2)

Key Documents

AB1555P

Sigma-Aldrich

Anti-NMDAR2A Antibody

Chemicon®, from rabbit

Synonym(s):

N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, NMDA receptor subtype 2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse, fish

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

General description

NMDA (N-methyl-D-aspartate) receptors are a class of ionotropic glutamate receptors. The NMDAR channel has been shown to be involved in long term potentiation, an activity dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D).

Specificity

NMDAR2A. By Western blot it recognizes a 180 kDa band in rat brain membranes. Does not react with NMDAR2B or NMDAR2C. Immunolabeling is blocked by preadsorption of antibody with the immunogen.

Immunogen

C-Terminal Fusion Protein of rat NMDAR2A, aa 1253-1391.
Epitope: C-terminus

Application

Anti-NMDAR2A Antibody is an antibody against NMDAR2A for use in IH, IP & WB.
Immunoprecipitation:
3 μg of a previous lot was used to (under appropriate conditions) quantitatively immunoprecipitate all NMDAR2A in 200 μg of rat brain.

Immunohistochemistry:
A previous lot of this antibody was used at 1:1,000-1:2,000 dilution.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Routinely evaluated by Western Blot on Rat Brain Microsomal lysates.

Western Blot Analysis: 1:500 dilution of this lot detected NMDAR2A on 10 μg of Rat Brain Microsomal lysates.

Target description

180 kDa

Physical form

ImmunoAffinity Purified
Purified rabbit polyclonal lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50 μL of PBS. Contains no preservative.

Storage and Stability

Lyophilized material stable for 1 year at -20°C from date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up 6 months.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
NMDAR2A is present in high concentrations in the hippocampus.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Co-Application of Corticosterone and Growth Hormone Upregulates NR2B Protein and Increases the NR2B:NR2A Ratio and Synaptic Transmission in the Hippocampus.
Mahmoud, GS; Amer, AS
Sultan Qaboos University medical journal null
Clàudia Cerveró et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 13(1), 198-216 (2015-11-20)
Spinal muscular atrophy (SMA) is a genetic neuromuscular disorder characterized by spinal and brainstem motor neuron (MN) loss and skeletal muscle paralysis. Currently, there is no effective treatment other than supportive care to ameliorate the quality of life of patients
Jennifer D Cooke et al.
Psychopharmacology, 231(23), 4429-4441 (2014-05-02)
Monoamine reuptake inhibitors can stimulate expression of brain-derived neurotrophic factor (BDNF) and alter long-term potentiation (LTP), a widely used model for the synaptic mechanisms that underlie memory formation. BDNF expression is upregulated during LTP, and BDNF in turn positively modulates
Lei-Lei Liu et al.
Frontiers in cellular neuroscience, 11, 279-279 (2017-09-30)
Recently, a line of evidence has demonstrated that the vertebrate retina possesses a novel retrograde signaling pathway. In this pathway, phototransduction is initiated by the photopigment melanopsin, which is expressed in a small population of retinal ganglion cells. These ganglion
Caroline Corbel et al.
Neural development, 10, 20-20 (2015-09-19)
N-methyl-D-aspartate receptors (NMDARs) are a subtype of ionotropic glutamate receptors and are expressed throughout the central nervous system (CNS). Their activity is required for excitatory synaptic transmission, the developmental refinement of neural circuits and for the expression of many forms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service