Skip to Content
Merck
All Photos(3)

Key Documents

07-632

Sigma-Aldrich

Anti-NR2A Antibody

Upstate®, from rabbit

Synonym(s):

N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor channe, subunit epsilon-1, N-methyl-D-aspartate receptor subunit 2A, NMDA receptor subtype 2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

General description

NR2 or glutamate receptor, ionotropic, N-methyl D-aspartate 2 (NMDA2; NMDAR2) along with the NR1 subunit forms a heterodimer that make up the NMDA receptor. By alternative splicing there are various isoforms of NR2 classified as NR2A through D. The NMDA receptor (NMDAR) is characterized as a ionotropic receptor for glutamate. As the name suggests, (NMDA (N-methyl D-aspartate) is an agonist for the receptor. A distinct characteristic of the NMDA receptor is that it is both ligand-gated and voltage-dependent based on its sensitivity to magnesium. The NR1 subunit binds glycine while the NR2 subunit binds glutamine for activation. Activated NMDA receptors result in the opening of an ion channel that is nonselective to cations. The result is the flow of Na+ and small amounts of Ca2+ ions into the cell and K+ out of the cell. Calcium flux by the NMDARs has been implicated in synaptic plasticity, a mechanism in the cell for learning and memory.

Specificity

Recognizes NR2A, MW ~170 kDa.

Immunogen

6His-tagged fusion protein corresponding to amino acids 1265-1464 of mouse NMDA receptor 2A (NR2A).

Application

Detect NR2A using this Anti-NR2A Antibody validated for use in IP & WB.
Immunoprecipitation:
4 µg of a previous lot immuno-precipitated NR2A from rat brain hippocampus membranes and rat brain microsomal preparations.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Evaluated by western blot in rat brain microsomal preparations (Catalog # 12-144).

Western Blot Analysis:
0.5-2 µg/mL of this antibody detected NR2A in rat brain microsomal preparations (Catalog # 12-144).

Target description

~170 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 70% storage buffer 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Brain tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human immunodeficiency virus (HIV)-induced neurotoxicity: roles for the NMDA receptor subtypes.
O'Donnell, LA; Agrawal, A; Jordan-Sciutto, KL; Dichter, MA; Lynch, DR; Kolson, DL
The Journal of Neuroscience null
Autism-linked neuroligin-3 R451C mutation differentially alters hippocampal and cortical synaptic function.
Etherton M, Foldy C, Sharma M, Tabuchi K, Liu X, Shamloo M, Malenka RC, Sudhof TC
Proceedings of the National Academy of Sciences of the USA null
GluN2B subunit deletion reveals key role in acute and chronic ethanol sensitivity of glutamate synapses in bed nucleus of the stria terminalis.
Wills, TA; Klug, JR; Silberman, Y; Baucum, AJ; Weitlauf, C; Colbran, RJ; Delpire, E; Winder, DG
Proceedings of the National Academy of Sciences of the USA null
April P Neal et al.
Neurotoxicology, 32(2), 281-289 (2011-01-05)
N-methyl-D-aspartate receptor (NMDAR) ontogeny and subunit expression are altered during developmental lead (Pb²+) exposure. However, it is unknown whether these changes occur at the synaptic or cellular level. Synaptic and extra-synaptic NMDARs have distinct cellular roles, thus, the effects of
Ilse Delint-Ramirez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(24), 8162-8170 (2010-06-18)
Lipid rafts are dynamic membrane microdomains enriched in cholesterol and sphingolipids involved in the compartmentalization of signaling pathways, trafficking and sorting of proteins. At synapses, the glutamatergic NMDA receptor and its cytoplasmic scaffold protein PSD-95 move between postsynaptic density (PSD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service