Skip to Content
Merck
All Photos(1)

Key Documents

1.46383

Millipore

MacConkey Broth

ready-to-use, bottle volume 100 mL , filling volume

Synonym(s):

Broth Medium G, MacC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
NACRES:
NA.74

Agency

EP 2.6.13
USP 62

Quality Level

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type wide neck
ready-to-use

packaging

bottle of
single packed of (100 ml in 275 ml wide mouth bottle (6 bottles per box))

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

275 mL

bottle volume

100 mL , filling volume

pH

7.3

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

Escherichia coli
coliforms
enterobacteriaceae

Looking for similar products? Visit Product Comparison Guide

General description

The MacConkey Broth supports the growth of most Gram-negative bacteria, while it inhibits the growth of Gram-positive bacteria due to the presence of bile salts. Bacteria that degrade lactose to acids induce a color change in the medium from violet to yellow. The change in color is indicated by the pH indicator bromocresol purple, which shows reduced inhibition compared to neutral red.

Application

MacConkey Broth is designed for the enrichment of Enterobacteriaceae and the detection of Escherichia coli in pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service