Skip to Content
Merck
All Photos(1)

Key Documents

1.46074

Millipore

HEIMPLATE R3A Agar - LI 30ml

For subculturing microorganisms found in pharmaceutical or potable water, sterile, ready-to-use

Synonym(s):

R-3A medium for long incubation, Reasoner′s 3A agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

pkg of 20 plates per box (90 mm settle plate) for long incubation (LI)

storage condition

protect from light (Store dry and tightly closed)

parameter

30 mL filling volume

technique(s)

microbiological culture: suitable

color

off-white

pH

7.2 ( in H2O)

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

heterotrophic bacteria

General description

R3A medium is a suitable subculture medium for organisms that either fail to grow on poorly nutritive R2A Agar or that are suppressed by other microorganisms on highly nutritive media (such as Tryptic Soy Agar or Plate Count Agar). Upon establishing the growth of microorganisms on R2A as a primary media, the higher nutritive R3A yields colonies, which are more suitable for biochemical identification.

Application

R3A Agar is used to subculture microorganisms found in pharmaceutical or potable water as well as for the primary isolation of aquatic microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service