Skip to Content
Merck
All Photos(1)

Key Documents

M-039

Supelco

Meprobamate solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4
CAS Number:
Molecular Weight:
218.25
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CCCC(C)(COC(N)=O)COC(N)=O

InChI

1S/C9H18N2O4/c1-3-4-9(2,5-14-7(10)12)6-15-8(11)13/h3-6H2,1-2H3,(H2,10,12)(H2,11,13)

InChI key

NPPQSCRMBWNHMW-UHFFFAOYSA-N

General description

Meprobamate is marketed as an anti-anxiety drug and muscle relaxant under the trade names Milltown, Meprospan, and Equanil. This Certified Spiking Solution® is suitable for GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iain M McIntyre et al.
Journal of analytical toxicology, 36(3), 177-181 (2012-03-16)
Carisoprodol is a therapeutic and occasionally abused centrally acting muscle relaxant. We compare central blood and liver concentrations of carisoprodol and the metabolite meprobamate to concentrations in peripheral blood in 11 medical examiner cases. Specimens were initially screened for alcohol
Arun Kumar et al.
Regulatory toxicology and pharmacology : RTP, 57(2-3), 146-156 (2010-02-16)
This study presents a step-wise development of a quantitative pharmaceutical risk assessment (QPhRA, hereafter) framework, including Monte Carlo uncertainty analysis for meprobamate, carbamazepine, and phenytoin during (1) accidental exposures of stream water and fish consumption and (2) direct ingestion of
Marina Del Fiacco et al.
Journal of neurology, neurosurgery, and psychiatry, 86(4), 393-397 (2014-10-08)
The transient receptor potential vanilloid type-1 receptor (TRPV1) and the neuropeptides calcitonin gene-related peptide (CGRP) and substance P (SP) appear to be differently involved in migraine pain. A role of neurovascular scalp structures is also suggested by several data. We
Bonnie V Laws et al.
The Science of the total environment, 409(6), 1087-1094 (2011-01-08)
The attenuation of a diverse suite of contaminants of emerging concern (CECs) and bulk water quality changes was evaluated at a surface-spreading aquifer recharge operation across a detailed subsurface profile (9 locations), representing both short- and long-travel times (10 h
Marie Orre et al.
Neurobiology of aging, 35(12), 2746-2760 (2014-07-09)
Reactive astrocytes and microglia are associated with amyloid plaques in Alzheimer's disease (AD). Yet, not much is known about the molecular alterations underlying this reactive phenotype. To get an insight into the molecular changes underlying AD induced astrocyte and microglia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service