Skip to Content
Merck
All Photos(1)

Key Documents

870439C

Avanti

C18:2 anandamide phosphate

Avanti Research - A Croda Brand 870439C

Synonym(s):

linoleoyl ethanolamide phosphate (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37NO5PNa
CAS Number:
Molecular Weight:
425.48
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (870439C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870439C

concentration

1 mg/mL (870439C-1mg)

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\C/C=C\CCCCC)NCCOP(O)([O-])=O.[Na+]

InChI key

PVAGBVTWXZTXFO-NBTZWHCOSA-M

General description

Anandamide is produced by the enzymatic hydrolysis of acyl phosphatidyl ethanolamides by phospholipase D.

Biochem/physiol Actions

Anandamide acts as a ligand for cannabinoid receptors. Anandamide possesses anti-inflammatory and analgesic properties. Its effect is regulated by cannabinoid CB1 and CB2 receptors.

Packaging

5 mL Amber Screw Cap Bottle with Foil Bag Argon Overseal (870439C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of linoleoyl ethanolamide
Wang X, et al.
Journal of Oleo Science, 62(6), 427-433 (2013)
Ocular hypotensive effect of oral palmitoyl-ethanolamide: a clinical trial
Gagliano C, et al.
Investigative Ophthalmology & Visual Science, 52(9), 6096-6100 (2011)
Protective role of palmitoylethanolamide in contact allergic dermatitis
Petrosino S, et al.
Allergy, 65(6), 698-711 (2010)
Anandamide receptors
Di Marzo V, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 66(2-3), 377-391 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service