Skip to Content
Merck
All Photos(1)

Key Documents

870431P

Avanti

C17:1 anandamide

Avanti Research - A Croda Brand 870431P, powder

Synonym(s):

10Z-heptadecenoylethanolamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H37NO2
CAS Number:
Molecular Weight:
311.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870431P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870431P

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCCCC/C=C\CCCCCC)NCCO

InChI key

IGADVTJDHNVILM-FPLPWBNLSA-N

General description

C17:1 anandamide, also known as 10Z-heptadecenoylethanolamide, is a N-acyl ethanolamide derivative of heptadecanoic acid.

Application

C17:1 anandamide or 10Z-heptadecenoylethanolamide has been used as an internal standard in mass spectrometry analysis for the measurement of 2-arachidonoyl glycerol (2-AG) in brain tissue. It has also been used as a substrate for N-acylethanolamine acid amidase (NAAA) enzyme.

Packaging

5 mL Clear Glass Sealed Ampule (870431P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoki Nakamura et al.
eLife, 8 (2019-10-10)
Cocaine is an addictive drug that acts in brain reward areas. Recent evidence suggests that cocaine stimulates synthesis of the endocannabinoid 2-arachidonoylglycerol (2-AG) in midbrain, increasing dopamine neuron activity via disinhibition. Although a mechanism for cocaine-stimulated 2-AG synthesis is known
Oscar Sasso et al.
Pain, 154(3), 350-360 (2012-12-12)
Fatty acid ethanolamides (FAEs), which include palmitoylethanolamide (PEA) and oleoylethanolamide (OEA), are endogenous agonists of peroxisome proliferator-activated receptor-α (PPAR-α) and important regulators of the inflammatory response. They are degraded in macrophages by the lysosomal cysteine amidase, N-acylethanolamine acid amidase (NAAA).
Natsuo Ueda et al.
Biochimica et biophysica acta, 1801(12), 1274-1285 (2010-08-26)
Ethanolamides of different long-chain fatty acids constitute a class of endogenous lipid molecules generally called N-acylethanolamines (NAEs). They contain N-arachidonoylethanolamine (anandamide), N-palmitoylethanolamine, and N-oleoylethanolamine, which receive considerable attention because of their actions as an endogenous cannabinoid receptor ligand (endocannabinoid), an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service