Skip to Content
Merck
All Photos(3)

Key Documents

840059P

Avanti

Hydro Egg PC

L-α-phosphatidylcholine, hydrogenated (Egg, Chicken), powder

Synonym(s):

Hydrogenated Egg Lecithin; HEL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (840059P-200mg)
pkg of 1 × 25 mg (840059P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840059P

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H84NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3/t40-/m1/s1

InChI key

PZNPLUBHRSSFHT-RRHRGVEJSA-N

General description

Hydrogenated egg PC (HPC) is made up of saturated fatty acids.

Application

Hydro Egg PC (hydrogenated egg lecithin) may be used:
  • in vesicles/liposomes for permeability studies to study the effect of MccJ25 on the permeability of phospholipid vesicles
  • to prepare small unilamellar vesicles (SUV) and multilamellar vesicles (MLV)
  • to prepare giant unilamellar vesicles
  • to prepare lipid emulsion to study the effects of phosphatidylcholine (PC) from different sources on intestinal absorption of cholesterol

Biochem/physiol Actions

Hydrogenated egg PC/ L-α-phosphatidylcholine (HPC) is a physiological surfactant.

Packaging

5 mL Amber Glass Screw Cap Vial (840059P-200mg)
5 mL Amber Glass Screw Cap Vial (840059P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abeer A W Elzainy et al.
The AAPS journal, 6(3), e18-e18 (2005-03-12)
Cetirizine, an effective, minimally sedating, second-generation H1-antihistamine is widely used orally to treat allergic skin disorders. This study was performed to assess the peripheral H1-antihistaminic activity and extent of systemic absorption of cetirizine from liposomes applied to the skin. Cetirizine
Morphological transitions of vesicles induced by AC electric fields Supplementary material
Aranda S, et al.
Biophysical Journal (2008)
A J Fillery-Travis et al.
Biophysical chemistry, 54(3), 253-260 (1995-05-01)
The emulsion phase formed within the stomach and duodenum during digestion of a fatty meal has been modelled using two physiological surfactants, the phospholipid L-alpha-phosphatidylcholine (PC) and the bile salt sodium taurocholate (NaT). Upon dilution of the phospholipid stabilised emulsions
Y Jiang et al.
The Journal of nutrition, 131(9), 2358-2363 (2001-09-05)
This study was conducted to determine the effects of phosphatidylcholine (PC) from different sources on intestinal absorption of cholesterol. Male Sprague-Dawley rats were fed an AIN-93G diet containing soybean oil for 4 wk. Each rat with lymph cannula was infused
Griselda R Borthiry et al.
Chemistry & biodiversity, 5(8), 1545-1557 (2008-08-30)
Chromium (Cr) is a cytotoxic metal that can be associated with a variety of types of DNA damage, including Cr-DNA adducts and strand breaks. Prior studies with purified human cytochrome b(5) and NADPH:P450 reductase in reconstituted proteoliposomes (PLs) demonstrated rapid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service