Skip to Content
Merck
All Photos(4)

Key Documents

441601G

Avanti

L-α-Phosphatidylcholine

95% (Soy), Avanti Polar Lipids

Synonym(s):

L-α-phosphatidylcholine (95%) (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Product Name

Soy PC (95%), Avanti Research - A Croda Brand

Assay

>95% (TLC)

form

granular

packaging

pkg of 1 × 100 g (441601G-100g)
pkg of 1 × 50 g (441601G-50g)
pkg of 1 × 500 g (441601G-500g)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

This Natural Lipid is a mixture and the structure shown is only representative of one possible structure present in the product. Refer to chart for average fatty acid distribution.

Application

Soy PC (95%) has been used as a supplement for dorsal root ganglia (DRGs) culture. It has also been used to determine the partitioning of a guest N-myristoylated methyl glycine (MrG) dipeptide.

Packaging

125 mL Amber Wide Mouth Glass Bottle with Screw Cap (441601G-50g)
1L Amber Wide Mouth Glass Bottle with Screw Cap (441601G-500g)
250 mL Amber Wide Mouth Glass Bottle with Screw Cap (441601G-100g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partitioning of an anchor dipeptide in a phospholipid membrane
Volkov VV and Righini R
The Journal of Physical Chemistry B, 113(50), 16246-16250 (2009)
T N Pashirova et al.
International journal of pharmaceutics, 575, 118953-118953 (2019-12-18)
In this study, we report the relationship between structure, self-assembly behavior and antimicrobial activity of multicationic gemini surfactants and their successful use as stabilizers of a new liposomal formulation for transdermal drug delivery. New surfactants containing natural moiety 1,4-diazabicyclo[2.2.2]octane with
Haiyang Zhang et al.
Journal of agricultural and food chemistry, 67(47), 13119-13126 (2019-11-07)
Phospholipids have been widely used in food, medicine, cosmetics, and other fields because of their unique chemical structure and healthcare functions. Phospholipase D (PLD) is a key biocatalyst for the biotransformation of phospholipids. Here, an autodisplay expression system was constructed
Jay C Fernandes et al.
Langmuir : the ACS journal of surfaces and colloids, 35(50), 16630-16639 (2019-12-06)
Dispersants, used in the mitigation of oil spills, are mixtures of amphiphilic molecules (surfactants) dissolved in a solvent. The recent large-scale use of dispersants has raised environmental concerns regarding the safety of these materials. In response to these concerns, our
Tarek A Ahmed
BMC pharmacology & toxicology, 21(1), 14-14 (2020-02-23)
Rosuvastatin (RSV) is a poorly water-soluble drug that has an absolute oral bioavailability of only 20%. The aim of this work was to prepare a positively charged chitosan coated flexible lipid-based vesicles (chitosomes) and compare their characteristics to the corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service