810283P
Avanti
TopFluor™ PS
Avanti Research™ - A Croda Brand 810283P, powder
Synonym(s):
1-palmitoyl-2-(dipyrrometheneboron difluoride)undecanoyl-sn-glycero-3-phospho-L-serine (ammonium salt)
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C46H80BF2N4O10P
CAS Number:
Molecular Weight:
928.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (810283P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 810283P
shipped in
dry ice
storage temp.
−20°C
General description
TopFluor™, a fluorescent molecule carries dipyrromethene boron difluoride moiety. It is linked to several lipid molecules by means of covalent attachment. Phosphoserine/phosphatidylserine (PS) is an endogenous phospholipid, that is found in all polyanionic macromolecules.
Application
TopFluor™ PS may be used in liposome generation and as a synthetic fluorescent phosphoserine (PS) analogue to study PS distribution and dynamics in mouse embryonic fibroblasts (MEFs). It has also been used in the preparation of donor liposomes.
Biochem/physiol Actions
Phosphoserine (PS) can be used as a template for the recognition of cancer antigen 125 (CA 125). It is very essential for healthy nerve cell membranes and myelin. PS actively participates in the locomotor functions, like rapid reactions and reflexes.
Packaging
5 mL Amber Glass Screw Cap Vial (810283P-1mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phosphatidylserine dynamics in cellular membranes
Kay JG, et al.
Molecular Biology of the Cell, 23(11), 2198-2212 (2012)
Deficient endoplasmic reticulum-mitochondrial phosphatidylserine transfer causes liver disease
Hernandez-Alvarez MI, et al.
Cell, 177(4), 881-895 (2019)
Lysosomal integral membrane protein-2 as a phospholipid receptor revealed by biophysical and cellular studies
Conrad KS, et al.
Nature Communications, 8(1908), 1-13 (2017)
Adsorption of O-phospho-L-serine and L-serine onto poorly crystalline apatite
Benaziz L, et al.
Journal of Colloid and Interface Science, 238(1), 48-53 (2001)
Investigation of the biophysical properties of a fluorescently modified ceramide-1-phosphate
Shirey CM, et al.
Chemistry and Physics of Lipids, 200, 32-32 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service