Skip to Content
Merck
All Photos(2)

Key Documents

810251P

Avanti

NBD-6 Cholesterol

Avanti Research - A Croda Brand 810251P, powder

Synonym(s):

5-cholesten-3β-ol 6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]caproate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H58N4O5
CAS Number:
Molecular Weight:
662.90
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810251P-1mg)
pkg of 1 × 5 mg (810251P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand 810251P

shipped in

dry ice

storage temp.

−20°C

General description

NBD-6 Cholesterol is a fluorescent analog of the lipoprotein compound cholesterol, with NBD (2-(4-nitro-2,1,3-benzoxadiazol-7-yl) aminoethyl) being the fluorescent group.

Application

NBD-6 Cholesterol has been used for NBD-cholesterol binding assay, to study the binding of fluorescent NBD cholesterol in the binding pocket concerning ASTER domains.

Biochem/physiol Actions

Cholesterol is produced from acetyl-CoA by a complex multistep mevalonate pathway. In mammals, liver is the main site of de novo cholesterol synthesis contributing to almost 80% of total cholesterol synthesis. Cholesterol and its metabolites, along with its precursors, play important roles in cellular membrane physiology. They also have roles in reproductive biology, dietary nutrient absorption, stress responses, salt and water balance and calcium metabolism.

Packaging

5 mL Amber Glass Screw Cap Vial (810299P-1MG)
5 mL Amber Glass Screw Cap Vial (810251P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fluorescent and Luminescent Probes for Biological Activity (1999)
Molecular Nutrition And Food Research, 277-290 (2016)
Cholesterol in health and disease.
Ira Tabas
The Journal of clinical investigation, 110(5), 583-590 (2002-09-05)
Pedro M R Cruz et al.
Frontiers in pharmacology, 4, 119-119 (2013-10-05)
While the unique metabolic activities of malignant tissues as potential targets for cancer therapeutics has been the subject of several recent reviews, the role of cholesterol metabolism in this context is yet to be fully explored. Cholesterol is an essential
Jaspreet Sandhu et al.
Cell, 175(2), 514-529 (2018-09-18)
The mechanisms underlying sterol transport in mammalian cells are poorly understood. In particular, how cholesterol internalized from HDL is made available to the cell for storage or modification is unknown. Here, we describe three ER-resident proteins (Aster-A, -B, -C) that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service