Skip to Content
Merck
All Photos(3)

Key Documents

841148P

Avanti

Soy PG

Avanti Research - A Croda Brand 841148P, powder

Synonym(s):

L-α-phosphatidylglycerol (Soy) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (841148P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 841148P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C40H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,37-38,41-42H,3-10,12,14-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b13-11-,18-17-;/t37?,38-;/m1./s1

InChI key

JBOGSUDWCXWRFD-YBBFZHODSA-M

General description

Soy PG is a phosphatidylglycerol (PG) mixture containing greater proportion of polyunsaturated fatty acids.

Application

Soy PG or L-α-phosphatidylglycerol (Soy) (sodium salt) might be used:
  • to study the interaction between phosphatidylglycerol (PG) and recombinant protein, AtROP6 (Rho-like GTPases), by comparing the classical protein lipid overlay (CPLO) and modified PLO (MPLO) assays
  • in liposome sedimentation assay, microscale thermophoresis (MST) assay, ROP6 activity assay and PG profile analysis, to study the role of PG in activation of ROP6 GTPase in Arabidopsis
  • in liposomes to study its effect on mouse keratinocyte proliferation

Biochem/physiol Actions

Soy PG is known to effectively prevent keratinocyte proliferation. Thus, it can treat keratinocyte hyperproliferation.

Packaging

5 mL Clear Glass Sealed Ampule (841148P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Xiuli Han et al.
Frontiers in plant science, 9, 347-347 (2018-03-31)
Plant Rho-like GTPases (ROPs) are switch-like proteins which play essential roles in controlling cell polarity development and cellular activities. ROPs are regulated by many factors, such as auxin, light, and RopGEFs and RopGAPs proteins. However, it has not been reported
Ding Xie et al.
The Journal of pharmacology and experimental therapeutics, 366(1), 1-8 (2018-04-27)
We have previously shown that phosphatidylglycerol (PG) regulates the function of keratinocytes, the predominant cells that compose the epidermis, inhibiting the proliferation of rapidly dividing keratinocytes. In particular, soy PG, a PG mixture with a high proportion of polyunsaturated fatty
Xiuli Han et al.
Plant methods, 16, 33-33 (2020-03-14)
Lipids perform multiple functions in the cell, and lipid-protein interactions play a key role in metabolism. Although various techniques have been developed to study lipid-protein interactions, the interacting protein partners that bind to most lipids remain unknown. The protein lipid
Ding Xie et al.
PloS one, 9(9), e107119-e107119 (2014-09-19)
We have previously shown that liposomes composed of egg-derived phosphatidylglycerol (PG), with a mixed fatty acid composition (comprising mainly palmitate and oleate), inhibit the proliferation and promote the differentiation of rapidly dividing keratinocytes, and stimulate the growth of slowly proliferating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service