Skip to Content
Merck
All Photos(1)

Key Documents

W500615

Sigma-Aldrich

Diacetin

Synonym(s):

Glycerol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COOCH2)2CHOH
CAS Number:
Molecular Weight:
176.17
Beilstein:
1706903
EC Number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

vapor density

6.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

refractive index

n20/D 1.440 (lit.)

density

1.17 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

alcohol

SMILES string

CC(=O)OCC(O)COC(C)=O.CC(=O)OCC(CO)OC(C)=O

InChI

1S/2C7H12O5/c1-5(8)11-3-7(10)4-12-6(2)9;1-5(9)11-4-7(3-8)12-6(2)10/h7,10H,3-4H2,1-2H3;7-8H,3-4H2,1-2H3

InChI key

TWSUHSVPDUPKDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diacetin is a volatile acetylated glycerol used as a flavoring agent in e-cigarettes. It is also found in floral oil secreting plants, where it is reported to act as a signal for oil-collecting bees to locate these plants, thus playing a key role in pollination.

Application


  • Organ-specific volatiles from Sonoran desert Krameria flowers as potential signals for oil-collecting bees.: This study identifies specific volatile compounds, including Diacetin, emitted by Krameria flowers. These volatiles act as signals for oil-collecting bees, suggesting a specialized ecological interaction (Balbuena et al., 2024, Phytochemistry).

  • Techno-Economic Assessment and Sensitivity Analysis of Glycerol Valorization to Biofuel Additives via Esterification.: This paper examines the economic viability of producing biofuel additives, including Diacetin, through the esterification of glycerol, highlighting the process′s potential for cost-effective biofuel production (Pandit et al., 2023, Industrial & Engineering Chemistry Research).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diacetin, a reliable cue and private communication channel in a specialized pollination system.
Schaffler I, et al.
Scientific Reports, 5, 12779-12779 (2105)
Comparison between electronic cigarette refill liquid and nicotine on metabolic parameters in rats.
Golli NE, et al.
Life Sciences, 146, 131-138 (2016)
Miguel Castañeda-Zárate et al.
Current biology : CB, 31(1), 238-246 (2020-11-07)
During the evolutionary history of flowering plants, transitions between pollinator groups (pollinator shifts) have been frequent,1 and contributed to the spectacular radiation of angiosperms.2 Although the evolution of floral traits during pollinator shifts has been studied in real time under
Irmgard Schäffler et al.
Scientific reports, 5, 12779-12779 (2015-08-08)
The interaction between floral oil secreting plants and oil-collecting bees is one of the most specialized of all pollination mutualisms. Yet, the specific stimuli used by the bees to locate their host flowers have remained elusive. This study identifies diacetin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service