
Diacetin

Synonym(s):
Glycerol diacetate
Linear Formula:
(CH3COOCH2)2CHOH
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing