Skip to Content
Merck
All Photos(3)

Key Documents

912042

Sigma-Aldrich

joYPhos

Umicore

Synonym(s):

(CyYPhos)(Ph)PCy2, Tricyclohexyl((dicyclohexyl-phosphanyl)(phenyl)methylene)-phosphane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H60P2
CAS Number:
Molecular Weight:
566.82
UNSPSC Code:
12352001
NACRES:
NA.22

Quality Level

form

powder

reaction suitability

reagent type: ligand

mp

205-214 °C

functional group

phosphine

General description

joYPhos is an ylide-functionalized phosphine ligand developed in the lab of Prof. V. Gessner with demonstrated uses in Pd-catalyzed cross coupling reactions, including the arylation of ketones and arylation of amines.

Application

The electron-rich and sterically demanding joYPhos ligand has been used in the gold(I)-catalyzed hydroamination of acetylene,citation and has shown to be effective in a range of Buchwald-Hartwig amination reactions. The strong electron-donor strength and sterically demanding nature of the ligand has been shown to increase the rate of formation of the catalytically active mono-phosphine palladium species, often leading to decreased reaction times or allowing the use of lower reaction temperatures.

Learn more about ylide-functionalized phosphines (YPhos)

Features and Benefits

Advantages of the joYPhos ligand over less electron rich ligand sources include, increased substrate scope in Buchwald-Hartwig amination reactions, including aryl chlorides, the use of more mild reaction conditions and improved activity in in C-N and C-C cross coupling reactions.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at www.pmc.umicore.com
Yphos is a trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Qiang Hu et al.
Organic letters, 21(18), 7558-7562 (2019-08-31)
Ylide-functionalized phosphine (YPhos) ligands allow the palladium-catalyzed α-arylation of alkyl ketones with aryl chlorides with record setting activity. Using a cyclohexyl-substituted YPhos ligand, a wide range of challenging ketone substrates was efficiently and selectively monoarylated under mild conditions. A newly
Ilja Rodstein et al.
The Journal of organic chemistry, 85(22), 14674-14683 (2020-09-11)
Ylide-substituted phosphines have been shown to be excellent ligands for C-N coupling reactions under mild reaction conditions. Here we report studies on the impact of the steric demand of the substituent in the ylide-backbone on the catalytic activity. Two new
Ilja Rodstein et al.
Chemical science, 13(45), 13552-13562 (2022-12-13)
The use of well-defined palladium(ii) complexes as precatalysts for C-X cross-coupling reactions has improved the use of palladium catalysts in organic synthesis including large-scale processes. Whereas sophisticated Pd(ii) precursors have been developed in the past years to facilitate catalyst activation
Zhiyong Hu et al.
Angewandte Chemie (International ed. in English), 60(12), 6778-6783 (2021-01-12)
The coupling of aryl chlorides with Reformatsky reagents is a desirable strategy for the construction of α-aryl esters but has so far been substantially limited in the substrate scope due to many challenges posed by various possible side reactions. This
Jens Tappen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(19), 4281-4288 (2020-01-24)
Palladium allyl, cinnamyl, and indenyl complexes with the ylide-substituted phosphines Cy3 P+ -C- (R)PCy2 (with R=Me (L1) or Ph (L2)) and Cy3 P+ -C- (Me)PtBu2 (L3) were prepared and applied as defined precatalysts in C-N coupling reactions. The complexes are

Articles

Explore innovative palladium-catalyzed coupling reactions with ylide-substituted phosphines. Learn about their impressive capabilities, enabling milder conditions and access to aryl chlorides.

Explore innovative palladium-catalyzed coupling reactions with ylide-substituted phosphines. Learn about their impressive capabilities, enabling milder conditions and access to aryl chlorides.

YPhos ligands enable efficient palladium-catalyzed coupling reactions under mild conditions, enhancing the synthesis of complex organic molecules.

Explore innovative palladium-catalyzed coupling reactions with ylide-substituted phosphines. Learn about their impressive capabilities, enabling milder conditions and access to aryl chlorides.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service