775037
N-Boc-O-tosylhydroxylamine
95%
Synonym(s):
N-Boc-O-tosyl hydroxylamine, tert-Butyltosyloxycarbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H17NO5S
CAS Number:
Molecular Weight:
287.33
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
functional group
amine
tosylate
storage temp.
2-8°C
SMILES string
Cc1ccc(cc1)S(=O)(=O)ONC(=O)OC(C)(C)C
InChI
1S/C12H17NO5S/c1-9-5-7-10(8-6-9)19(15,16)18-13-11(14)17-12(2,3)4/h5-8H,1-4H3,(H,13,14)
InChI key
WZDPZKPHVNFUKB-UHFFFAOYSA-N
Application
Electrophilic N-amination of aryl and alkyl amines.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Baburaj, T.;
Synlett, 1993-1996 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service