Skip to Content
Merck
All Photos(1)

Key Documents

226157

Sigma-Aldrich

N-Boc-hydroxylamine

≥98%

Synonym(s):

tert-Butyl N-hydroxycarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHOH
CAS Number:
Molecular Weight:
133.15
Beilstein:
1756546
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

mp

53-55 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NO

InChI

1S/C5H11NO3/c1-5(2,3)9-4(7)6-8/h8H,1-3H3,(H,6,7)

InChI key

DRDVJQOGFWAVLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Useful hydroxylamine derivatives for the synthesis of hydroxamic acids.
Tetrahedron Letters, 33(35), 5055-5058 (1992)
Tetrahedron Letters, 33, 5055-5055 (1992)
Brian S Bodnar et al.
The Journal of organic chemistry, 72(10), 3929-3932 (2007-04-14)
The addition of azides to acylnitroso hetero-Diels-Alder cycloadducts derived from cyclopentadiene affords exo-triazolines in excellent yield. The reaction is greatly affected by the level of alkene strain, while sterically demanding azides do not hinder the reaction. Conversion of the triazolines
Synthetic Communications, 22, 2579-2579 (1992)
M J Lee et al.
Journal of medicinal chemistry, 35(20), 3648-3652 (1992-10-02)
In the preceding paper, analogs of chlorpropamide with an OMe substituent on the sulfonamide nitrogen were shown to inhibit aldehyde dehydrogenase (AlDH), and it was postulated that these compounds were bioactivated by O-demethylation to release nitroxyl (HN = O, nitrosyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service