Skip to Content
Merck
All Photos(1)

Key Documents

486205

Sigma-Aldrich

Tetrachloroethylene-13C1

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C=13CCl2
CAS Number:
Molecular Weight:
166.83
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C

mass shift

M+1

SMILES string

Cl\C(Cl)=[13C](/Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6/i1+1

InChI key

CYTYCFOTNPOANT-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service