Skip to Content
Merck
All Photos(1)

Key Documents

02666

Supelco

Tetrachloroethylene

analytical standard

Synonym(s):

PCE, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Beilstein:
1361721
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

5.83 (vs air)

vapor pressure

13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

Assay

≥99.9% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.505 (lit.)
n20/D 1.506

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cl\C(Cl)=C(\Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrachloroethylene is a chlorinated ethylene compound commonly used as a dry cleaning and degreasing solvent.[1] It is a groundwater contaminant, which has adverse effects on human health due to its potential toxicity and carcinogenicity.[2]

Application

Tetrachloroethylene may be used as an extraction solvent for the determination of oils and grease in water samples using the Fourier-transform infrared spectroscopy technique.[3] It may be used as an analytical reference standard for the quantification of the analyte in biological fluids using headspace solid-phase micro extraction-capillary gas chromatography with electron capture detection.[4]

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Validation of a FT-IR method for the determination of oils and grease in water using tetrachloroethylene as the extraction solvent
Farmaki E, et al.
Desalination, 210(1-3), 52-60 (2007)
Tetrachloroethylene and trichloroethylene fatality: case report and simple headspace SPME-capillary gas chromatographic determination in tissues
Dehon B, et al.
Journal of Analytical Toxicology, 24(1), 22-26 (2000)
A History of the Production and Use of Carbon Tetrachloride, Tetrachloroethylene, Trichloroethylene and 1, 1, 1-Trichloroethane in the United States: Part 2--Trichloroethylene and 1, 1, 1-Trichloroethane.
Doherty RE.
Environmental Forensics, 1(2), 83-93 (2000)
T M Vogel et al.
Applied and environmental microbiology, 49(5), 1080-1083 (1985-05-01)
Tetrachloroethylene (PCE) and trichloroethylene (TCE), common industrial solvents, are among the most frequent contaminants found in groundwater supplies. Due to the potential toxicity and carcinogenicity of chlorinated ethylenes, knowledge about their transformation potential is important in evaluating their environmental fate.
Dusty R V Berggren et al.
Environmental science & technology, 47(4), 1879-1886 (2013-01-16)
Results are presented from a chemostat study where the reductive dehalogenation of PCE was evaluated in the absence and presence of sulfate. Two chemostats inoculated with the Point Mugu culture, which contains strains of Dehalococcoides mccartyi, were operated at a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service