Skip to Content
Merck
All Photos(1)

Key Documents

CRM47930

Supelco

QTM PAH Mix

certified reference material, 2000 μg/mL each component in dichloromethane, ampule of 1 mL

Synonym(s):

PAH Standard Mix for QTM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benz[a]anthracene
    Benzo[b]fluoranthene
    Benzo[ghi]perylene
    Benzo(a)pyrene
    2-Bromonaphthalene
    Chrysene
    Dibenz[a,h]anthracene
    Fluoranthene
    Fluorene
    Indeno[1,2,3-cd]pyrene
    Phenanthrene
    Pyrene
    Naphthalene
    See All (16)

Signal Word

Danger

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service