48249
EPA 525 PAH Mix B
certified reference material, 500 μg/mL each component in acetone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Agency
EPA 525,525.1,525.2
product line
TraceCERT®
CofA
current certificate can be downloaded
feature
standard type calibration
packaging
ampule of 1 mL
concentration
500 μg/mL each component in acetone
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-30°C
Application
EPA 525 PAH Mix B has been used as an internal standard in the extraction and determination of polycyclic aromatic hydrocarbons in water and milksamples using solid phase micro extraction technique (SPME) and gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
Acenaphthylene
Anthracene
Benz[a]anthracene
Benzo[b]fluoranthene
Benzo[k]fluoranthene
Benzo[ghi]perylene
Benzo[a]pyrene
Chrysene
Dibenz[a,h]anthracene
Fluorene
Indeno[1,2,3-cd]pyrene
Phenanthrene
Pyrene
See All (13)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Supplementary Hazards
Flash Point(F)
1.4 °F
Flash Point(C)
-17 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Experimental design for the optimization of the extraction conditions of polycyclic aromatic hydrocarbons in milk with a novel diethoxydiphenylsilane solid-phase microextraction fiber
Journal of Chromatography A, 1196, 41-45 (2008)
Innovative sol?gel coatings for solid-phase microextraction: development of fibers for the determination of polycyclic aromatic hydrocarbons at trace level in water
Journal of Chromatography A, 1196, 15-22 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service