Key Documents
Z351539
Eppendorf® epT.I.P.S. Biopur®, singles
pack of 1000 ea
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
manufacturer/tradename:
Eppendorf® 022492209
L:
53 mm
maximum volume:
200 μL
sterility:
sterile
feature:
ATP free
DNA free (Bacterial (E. Coli))
DNA free (Human)
DNase free
RNase free
barrier: no
pyrogen free
packaging:
pack of 1000 ea
material
colorless
description
PCR inhibitor free
sterility
sterile
feature
ATP free
DNA free (Bacterial (E. Coli))
DNA free (Human)
DNase free
RNase free
barrier: no
pyrogen free
packaging
pack of 1000 ea
manufacturer/tradename
Eppendorf® 022492209
L
53 mm
maximum volume
200 μL
Related Categories
General description
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Application
Eppendorfe® epT.I.P.S.® Biopur®, singles are suitable for the medical, food industry, molecular biology, pharmaceutical, and cell technology applications.
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)