Key Documents
Z640174
Eppendorf® epT.I.P.S. Biopur®, singles
volume 2-200 μL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
2—200 μL
sterility:
sterile
packaging:
pack of 100 (individually wrapped)
manufacturer/tradename:
Eppendorf® 0030 010.035
Eppendorf® 022491148
material
colorless
sterility
sterile
packaging
pack of 100 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030 010.035
Eppendorf® 022491148
volume measuring range
2-200 μL
L
53 mm
maximum volume
200 μL
volume
2—200 μL
General description
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)