Skip to Content
Merck
All Photos(2)

Key Documents

T9792

Sigma-Aldrich

6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt

Synonym(s):

Sodium 6-(p-toluidino)-2-naphthalenesulfonate, TNS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14NNaO3S
CAS Number:
Molecular Weight:
335.35
Beilstein:
6836595
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

storage temp.

2-8°C

Quality Level

SMILES string

[H]O[H].OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C17H15NO3S.Na/c1-12-2-6-15(7-3-12)18-16-8-4-14-11-17(22(19,20)21)9-5-13(14)10-16;/h2-11,18H,1H3,(H,19,20,21);/q;+1/p-1

InChI key

QZZGQOXRGFDEJP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent probe for the conformational state of proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jennifer Estendorfer et al.
Frontiers in plant science, 8, 930-930 (2017-07-07)
In this study, we investigated the impact of different land use intensities (LUI) on the root-associated microbiome of
Takuro Ito et al.
Molecular phylogenetics and evolution, 113, 9-22 (2017-04-26)
We explored the temporal and spatial diversification of the plant genus Sedum L. (Crassulaceae) in Taiwan based on molecular analysis of nrITS and cpDNA sequences from East Asian Sedum members. Our phylogenetic and ancestral area reconstruction analysis showed that Taiwanese
Christian Than et al.
Journal of electromyography and kinesiology : official journal of the International Society of Electrophysiological Kinesiology, 41, 60-65 (2018-05-18)
This study investigated test-retest reliability of mechanomyography (MMG) on lumbar paraspinal muscles. Healthy male and female subjects (mean ± standard deviation, 25 ± 9.4 years, BMI 21.8 ± 2.99, n = 34) were recruited. Two test sessions (one week apart) consisted of MMG (laser displacement sensor (LDS)) muscle evaluations
Tushar Issar et al.
Clinical neurophysiology : official journal of the International Federation of Clinical Neurophysiology, 129(5), 889-894 (2018-03-17)
To demonstrate construct validity of the Total Neuropathy Score (TNS) in assessing peripheral neuropathy in subjects with chronic kidney disease (CKD). 113 subjects with CKD and 40 matched controls were assessed for peripheral neuropathy using the TNS. An exploratory factor
Neshatul Haque et al.
Physical chemistry chemical physics : PCCP, 19(36), 24656-24666 (2017-09-01)
Characterization of different conformational states of proteins is essential to understand their stability and activity. Biophysical techniques aid in analysing these conformational states and molecular fluorescence is one of the most reliable and quickly accessible methods. Apart from the intrinsic

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service