
6-(p-Toluidino)-2-naphthalenesulfonic acid sodium salt

Synonym(s):
Sodium 6-(p-toluidino)-2-naphthalenesulfonate, TNS
Empirical Formula (Hill Notation):
C17H14NNaO3S
CAS Number:
Molecular Weight:
335.35
Products
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing