Skip to Content
Merck
All Photos(1)

Key Documents

SML0657

Sigma-Aldrich

N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride

≥98% (HPLC)

Synonym(s):

W 5 hydrochloride, W-5 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22N2O2S · HCl
CAS Number:
Molecular Weight:
342.88
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder or crystals

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

room temp

SMILES string

[S](=O)(=O)(NCCCCCCN)c1c2c(ccc1)cccc2.Cl

InChI

1S/C16H22N2O2S.ClH/c17-12-5-1-2-6-13-18-21(19,20)16-11-7-9-14-8-3-4-10-15(14)16;/h3-4,7-11,18H,1-2,5-6,12-13,17H2;1H

InChI key

HOCSVIGHWPLMFC-UHFFFAOYSA-N

Biochem/physiol Actions

N-(6-Aminohexyl)-1-naphthalenesulfonamide (W-5) is a cell permeable calmodulin antagonist that binds directly to the enzyme.
N-(6-Aminohexyl)-1-naphthalenesulfonamide (W-5) is a cell permeable calmodulin antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Im et al.
The Plant cell, 8(12), 2245-2253 (1996-12-01)
The Chlamydomonas reinhardtii nuclear gene gsa, which encodes the early chlorophyll biosynthetic enzyme glutamate 1-semialdehyde aminotransferase (GSAT), is specifically induced by blue light in cells synchronized in a 12-hr-light and 12-hr-dark regime. Light induction required the presence of a nitrogen
Deirdre C Lawe et al.
Molecular biology of the cell, 14(7), 2935-2945 (2003-07-15)
Ca2+ is an essential requirement in membrane fusion, acting through binding proteins such as calmodulin (CaM). Ca2+/CaM is required for early endosome fusion in vitro, however, the molecular basis for this requirement is unknown. An additional requirement for endosome fusion
Y Tanaka et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(8), 3244-3248 (1995-04-11)
The ryanodine receptor-like Ca2+ channel (RyRLC) is responsible for Ca2+ wave propagation and Ca2+ oscillations in certain nonmuscle cells by a Ca(2+)-induced Ca2+ release (CICR) mechanism. Cyclic ADP-ribose (cADPR), an enzymatic product derived from NAD+, is the only known endogenous
A Takahashi et al.
Pancreas, 21(3), 231-239 (2000-10-20)
The mouse intestinal neuroendocrine tumor cell line STC-1 secretes cholecystokinin (CCK) and other hormones. We investigated the role of Ca2+, calmodulin (CaM), and protein kinase C (PKC) in the regulation of CCK release from STC-1 cells. Phorbol 12-myristate 13-acetate (TPA)
K Ahmad et al.
Archives of andrology, 35(3), 187-208 (1995-11-01)
The role of Ca2+, calmodulin, and protein phosphatases on motility and hyperactivation of noncapacitated, capacitating, and detergent-permeabilized reactivated human sperm was examined. In noncapacitated sperm, W7 inhibited percent motility (%MOT), curvilinear velocity (VCL), amplitude of lateral head displacement (ALH), and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service