Skip to Content
Merck
All Photos(1)

Key Documents

422706

Sigma-Aldrich

KN-62

A cell-permeable, reversible, and selective inhibitor of CaM kinase II (Ki = 900 nM for rat brain CaM kinase II) that binds directly to the calmodulin binding site of the enzyme.

Synonym(s):

KN-62, 1-[N,O- bis-(5-Isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine, P2X Antagonist II, Purinergic Receptor P2X Antagonist II, 1-[N,O-bis-(5-Isoquinolinesulfonyl)-N-methyl-L-tyrosyl]-4-phenylpiperazine, P2X Antagonist II, Purinergic Receptor P2X Antagonist II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H35N5O6S2
CAS Number:
Molecular Weight:
721.84
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white

solubility

methanol: 5 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C38H35N5O6S2/c1-41(50(45,46)36-11-5-7-29-26-39-19-17-33(29)36)35(38(44)43-23-21-42(22-24-43)31-9-3-2-4-10-31)25-28-13-15-32(16-14-28)49-51(47,48)37-12-6-8-30-27-40-20-18-34(30)37/h2-20,26-27,35H,21-25H2,1H3

InChI key

RJVLFQBBRSMWHX-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and selective inhibitor of Ca2+/calmodulin-dependent protein kinase II (Ki = 900 nM for rat brain enzyme) that binds directly to the calmodulin binding site of the enzyme. Also inhibits the growth of K562 cells in a dose-dependent manner.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cam kinase 2
Product does not compete with ATP.
Reversible: yes
Target Ki: 900 nM for rat brain CaM kinase II

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffer just prior to use.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 4 months at 4°C.

Other Notes

Minami, H., et al. 1994. Biochem. Biophys. Res. Commun.199, 241.
Kato, M., et al. 1992. Neurosci. Lett.129, 47.
Ishii, A., et al. 1991. Biochem. Biophys. Res. Commun.176, 1051.
Ito, I., et al. 1991. Neurosci. Lett.121, 119.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilse Delint-Ramirez et al.
Molecular cell, 82(20), 3794-3809 (2022-10-08)
Neuronal activity induces topoisomerase IIβ (Top2B) to generate DNA double-strand breaks (DSBs) within the promoters of neuronal early response genes (ERGs) and facilitate their transcription, and yet, the mechanisms that control Top2B-mediated DSB formation are unknown. Here, we report that
Petra Muenzner et al.
Cell host & microbe, 27(5), 793-808 (2020-04-15)
Several pathogens suppress exfoliation, a key defense of epithelia against microbial colonization. Common among these pathogens, exemplified by Neisseria gonorrhoeae, is their ability to bind carcinoembryonic antigen-related cell adhesion molecules (CEACAMs). Gonococcal CEACAM engagement triggers the expression of CD105, which
Irma E Cisneros et al.
Frontiers in neurology, 11, 593146-593146 (2020-12-17)
Methamphetamine (METH) use, referred to as methamphetamine use disorder (MUD), results in neurocognitive decline, a characteristic shared with HIV-associated neurocognitive disorders (HAND). MUD exacerbates HAND partly through glutamate dysregulation. Astrocyte excitatory amino acid transporter (EAAT)-2 is responsible for >90% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service