Skip to Content
Merck
All Photos(4)

Documents

SAB5600227

Sigma-Aldrich

Anti-His-Tag antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM146, purified immunoglobulin

Synonym(s):

10 His epitope tag, 6 His epitope tag, HHHHHH tag, Polyhistidine Tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM146, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

all

technique(s)

flow cytometry: 0.5-2 μg/mL
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.1—1 μg/mL
immunoprecipitation (IP): 0.5-2 μg/mL
western blot: 0.1— 1 μg/mL

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

This antibody reacts to recombinant proteins containing the 6xHis-Tag or 10xHis-Tag fused to either the amino or carboxy terminus. No cross reactivity with other endogenous protein in mammalian or bacteria cells.

Immunogen

Mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis-Tag at the C-terminus.

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human inhalable antibody fragments neutralizing SARS-CoV-2 variants for COVID-19 therapy.
Minenkova, et al.
Molecular Therapy, 30, 1979-1993 (2022)
Tarah Elizabeth Trebino et al.
eLife, 12 (2023-12-27)
BRAF is a key member in the MAPK signaling pathway essential for cell growth, proliferation, and differentiation. Mutant BRAF is often the underlying cause of various types of cancer and mutant RAS, the upstream regulator of BRAF, is a driver
Harman Malhi et al.
Cell reports. Medicine, 3(6), 100658-100658 (2022-06-16)
Epstein-Barr virus (EBV) is a cancer-associated pathogen responsible for 165,000 deaths annually. EBV is also the etiological agent of infectious mononucleosis and is linked to multiple sclerosis and rheumatoid arthritis. Thus, an EBV vaccine would have a significant global health
Kristina R Edwards et al.
NPJ vaccines, 9(1), 120-120 (2024-06-27)
Epstein-Barr virus (EBV) is associated with several malignancies, neurodegenerative disorders and is the causative agent of infectious mononucleosis. A vaccine that prevents EBV-driven morbidity and mortality remains an unmet need. EBV is orally transmitted, infecting both B cells and epithelial
Kentaro Yumoto et al.
Antioxidants (Basel, Switzerland), 9(11) (2020-10-31)
Emphysema, a type of lung-destroying condition associated with chronic obstructive pulmonary disease (COPD), is an inflammatory lung disease mainly due to cigarette smoke exposure. As there is no curative therapy, prevention should be considered first by cessation of smoking to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service