SAB4600167
Monoclonal Anti-6X His IgG (H+L), CF™ 640R antibody produced in mouse
~1 mg/mL, clone 33D10.D2.G8, purified immunoglobulin
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
CF™ 640R conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
33D10.D2.G8, monoclonal
form
buffered aqueous solution
concentration
~1 mg/mL
technique(s)
indirect immunofluorescence: 1-10 μg/mL
isotype
IgG1κ
fluorescence
λex 642 nm; λem 662 nm
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Immunogen
Features and Benefits
Physical form
Preparation Note
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service