S4375
DL-Serine
≥98% (TLC)
Synonym(s):
(±)-2-Amino-3-hydroxypropionic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HOCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721405
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
DL-Serine, ≥98% (TLC)
Assay
≥98% (TLC)
form
powder
color
white
mp
240 °C (dec.) (lit.)
solubility
H2O: soluble
SMILES string
NC(CO)C(O)=O
InChI
1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)
InChI key
MTCFGRXMJLQNBG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
NMDA agonist acting at the glycine site; precursor of glycine by serine hydroxymethyltransferase
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Martin Picard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 118(9), 1161-1171 (2015-03-15)
The diaphragm is a unique skeletal muscle designed to be rhythmically active throughout life, such that its sustained inactivation by the medical intervention of mechanical ventilation (MV) represents an unanticipated physiological state in evolutionary terms. Within a short period after
Jacob T Bjerrum et al.
Inflammatory bowel diseases, 20(12), 2340-2352 (2014-10-31)
It is unknown why patients with extensive ulcerative colitis (UC) have a higher risk of colorectal cancer compared with patients with left-sided UC. This study characterizes the inflammatory processes in left-sided UC, pancolitis, and UC-associated dysplasia at the transcriptional level
K U Sales et al.
Oncogene, 34(3), 346-356 (2014-01-29)
The membrane-anchored serine protease, matriptase, is consistently dysregulated in a range of human carcinomas, and high matriptase activity correlates with poor prognosis. Furthermore, matriptase is unique among tumor-associated proteases in that epithelial stem cell expression of the protease suffices to
Marne C Hagemeijer et al.
Virology, 458-459, 125-135 (2014-06-15)
Coronaviruses replicate their genomes in association with rearranged cellular membranes. The coronavirus nonstructural integral membrane proteins (nsps) 3, 4 and 6, are key players in the formation of the rearranged membranes. Previously, we demonstrated that nsp3 and nsp4 interact and
Wei Wu et al.
Molecular cancer research : MCR, 12(12), 1717-1728 (2014-07-27)
Colorectal cancer metastasis is a major cause of mortality worldwide, which may only be controlled with novel methods limiting tumor dissemination and chemoresistance. High stathmin-1 (STMN1) expression was previously established as a hallmark of colorectal cancer progression and predictor of
Chromatograms
application for HPLCapplication for HPLCapplication for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service