Skip to Content
Merck
All Photos(3)

Key Documents

171824

Sigma-Aldrich

DL-Proline

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

(±)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80812
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

DL-Proline, ReagentPlus®, 99%

product line

ReagentPlus®

Assay

99%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

208 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)

InChI key

ONIBWKKTOPOVIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Proline, also known as 2-pyrrolidinylcarboxylic acid, is an amino acid commonly used in solution-phase peptide synthesis due to its ability to act as a turn inducer resulting from its restricted phi( ϕ) angle. [1]

Application

DL-Proline can be is used in the preparation of cis-aminoproline-alkene (cis-Apa)linker, which is further used as an intermediate compound for the synthesis of cyclic pseudopeptides via solution phase peptide synthesis. [2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

cis-Apa: A practical linker for the microwave-assisted preparation of cyclic pseudopeptides via RCM cyclative cleavage
A Baron
The Journal of Organic Chemistry, 76, 766-772 (2011)
Recent Progress in Solid-Phase Total Synthesis of Naturally Occurring Small Peptides
H Yan
Advanced Synthesis & Catalysis, 364, 1934-1961 (2022)
Antoinette Bugyei-Twum et al.
Cardiovascular diabetology, 13, 89-89 (2014-06-03)
Despite advances in the treatment of heart failure, mortality remains high, particularly in individuals with diabetes. Activated transforming growth factor beta (TGF-β) contributes to the pathogenesis of the fibrotic interstitium observed in diabetic cardiomyopathy. We hypothesized that high glucose enhances
Nina Maeshima et al.
The Journal of biological chemistry, 290(21), 13440-13453 (2015-04-04)
Lipid A in LPS activates innate immunity through the Toll-like receptor 4 (TLR4)-MD-2 complex on host cells. Variation in lipid A has significant consequences for TLR4 activation and thus may be a means by which Gram-negative bacteria modulate host immunity.
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service