Skip to Content
Merck
All Photos(2)

Documents

RAB0361

Sigma-Aldrich

Human MMP-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

MMP-1, Matrix metalloproteinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 24.69-18000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP1(4312)

General description

The Human MMP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MMP-1 pro and active forms in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MMP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human MMP-1 ELISA Kit has been used in ELISA assays.

Biochem/physiol Actions

Matrix metalloproteinases (MMPs) enzymes play a central role in the maintenance and remodeling of the extracellular matrix. Elevated expression of their activity, caused either by up-regulation of their expression or down-regulation of their cognate inhibitors, has been implicated in various degenerative disorders, including arthritis, cardiovascular disease, skeletal growth-plate disorders, and cancer metastasis. MMP-1 is a secreted collagenase with specificity toward Type I, II, III, VII, and X collagens, aggrecan, serpins, and α2-macroglobulin. It is overexpressed in lumbar intervertebral disc degeneration (IDD). It might also be associated with osteoarthritis. MMP-1 is linked with juvenile idiopathic arthritis. Genetic variations in MMP-1 can affect the susceptibility and severity of this disease.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of a matrix metalloproteinase-1 biosensor based on the surface plasmon resonance imaging technique
Tokarzewicz A, et al.
Analytical Methods : Advancing Methods and Applications, 8(34), 428-6435 (2016)
Influence of Matrix metalloproteinase 1 and 3 genetic variations on susceptibility and severity of juvenile idiopathic arthritis.
Abd-Allah SH, et. al.
IUBMB Life, 67, 934-942 (2015)
Expression profiles of MMP-1 and TIMP-1 in lumbar intervertebral disc degeneration.
Deng B, et. al.
Genetics and molecular research : GMR, 14, 19080-19086 (2015)
Regulation of matrix metalloproteinase expression in tumor invasion.
Westermarck J and Kahari VM
FEBS Journal, 13, 781-792 (1999)
MMP-2 functions as an early response protein in ovarian cancer metastasis.
Kenny HA, et. al.
Cell Cycle, 8, 683-688 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service