Merck
HomeProductsProtein BiologyImmunoassay Platform Solutions
Immunoassay Platform Solutions
Immunoassay Platform Solutions
Whether you need ultrasensitive detection from a SMC® assay, robust ELISAs, or rich multiplex profiling data using Luminex® xMAP® technology, we can help you get data you can trust out of each sample, with minimal repetition, so you can make the best next-step decisions. Learn more about our platforms including Single Molecule Counting (SMC®) instruments, kits, and reagents; our MILLIPLEX® kits for multiplex biomarker detection with Luminex® instruments; and our wide range of sandwich ELISAs.
ELISA Kits and Reagents
Enzyme-Linked Immunosorbent Assay (ELISA) Kits
An ELISA (Enzyme-Linked Immunosorbent Assay) is a multi-well plate-based immunoassay within which one of the assay components, typically an antibody or sample, is adsorbed onto a solid surface, in this case, a plate. Offering rapid, quantitative, and sensitive analyte detection at relatively low cost, ELISAs represent one of the simplest assay formats to perform. Furthermore, the ease of adapting an ELISA to a higher throughput screening method empowers researchers to test large sample numbers in a single run.
MILLIPLEX® Multiplex for Luminex® Immunoassays
Your time and samples are too precious to trust anything but the most highly verified MILLIPLEX® multiplex assays for the Luminex® platform. Robust assay performance data is included in every protocol ensuring the highest quality data.
MILLIPLEX® Multiplex for Luminex® Immunoassays
Explore More Categories
SMC® Immunoassays
SMC® Technology
Single Molecule Counting (SMC®) technology precisely quantifies molecules at previously undetectable levels.
View Products