Key Documents
RA817-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
A-8-17, aerosol-tight, to hold, 8 x 15 mL conical tubes, without adapter, without rubber mats
About This Item
UNSPSC Code:
41103909
feature
to hold 8 x 15 mL conical tubes
aerosol-tight
low-capacity swing bucket rotor
without adapter
without rubber mats
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639501 (NA)
Eppendorf® 5702700002 (EU)
parameter
2,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/5702RH
Related Categories
General description
Rotors to fit Eppendorf centrifuge models 5702, 5720 R, and 5720 RH
Legal Information
Eppendorf is a registered trademark of Eppendorf AG