Skip to Content
AW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges
Synonym(s):
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges
Products
Product Category
rotors (6)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP022639304
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
A-4-38, aerosol-tight, with 4 x 90 mL rectangular buckets
Expand
View Pricing
RA4384-EP
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
A-4-38, aerosol-tight, with 4 x 100 mL round buckets
Expand
View Pricing
RF353017C-EP
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
F-35-30-17, non aerosol tight, with 10 steel sleeves for 15 mL vessels, with 10 adapters for conical tubes and 10 rubber mats for round-bottom tubes
Expand
View Pricing
RA438-EP
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
A-4-38, aerosol-tight, without buckets, for round-bottom tubes only
Expand
View Pricing
RA817-EP
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
A-8-17, aerosol-tight, to hold, 8 x 15 mL conical tubes, without adapter, without rubber mats
Expand
View Pricing
EP022639404
Rotor for Eppendorf
®
5702/5702R/5702RH Centrifuges,
to hold, 30 x 15 mL vessels
Expand
View Pricing