Skip to Content
Merck
All Photos(1)

Documents

I8909

Sigma-Aldrich

Anti-Interleukin-1α antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-1α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable
western blot: 1-2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL1A(3552)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced by the activation of NF-κ B transcription factor in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signalling through IL-1 receptor type 1. A variety of cells such as macrophages, epithelial cells, fibroblasts, keratinocytes, monocytes and dendritic cells secrete IL-1α. Both IL-1α and IL-1β are important in acquired immunity, inflammation, response to infection and production of matrix metalloproteinases (MMPs). The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation. Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis
Anti-interleukin-1α specifically reacts with human IL-1α. It does not react with human IL-1β and mouse IL-1β or mouse IL-1α in neutralization reactions. It shows 5% cross reactivity with mouse IL-1α in immunoblotting applications.

Immunogen

purified, E. coli-derived recombinant human interleukin-1α.

Application

Anti-interleukin 1α antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. It is also suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-1 (IL-1) pathway
Weber A et al
Science Signaling, 3, p-p (2010)
S Z Ben-Sasson et al.
Cytokine, 56(1), 122-125 (2011-08-17)
IL-1 strikingly enhances antigen-driven responses of CD4 and CD8 T cells. It is substantially more effective than LPS and when added to a priming regime of antigen plus LPS, it strikingly enhances cell expansion. The effect is mediated by direct
Charles A Dinarello
Current opinion in pharmacology, 4(4), 378-385 (2004-07-15)
All biological agents currently used for reducing TNFalpha activity in disease are neutralization strategies; however, there are several strategies for reducing interleukin (IL)-1 activities: the IL-1 receptor antagonist (IL-1Ra), anti-IL-1beta monoclonal antibodies, the IL-1 Trap, IL-1 receptor type I antibodies
Emmanuel Contassot et al.
Swiss medical weekly, 142, w13590-w13590 (2012-06-02)
Interleukin 1, one of the first cytokines discovered in the 1980s, and a potent mediator of fever, pain and inflammation, is at present experiencing a revival in biology and medicine. Whereas the mechanism of activation and secretion of interleukin 1β
H E Barksby et al.
Clinical and experimental immunology, 149(2), 217-225 (2007-06-26)
Understanding cytokine immunobiology is central to the development of rational therapies for destructive inflammatory diseases such as rheumatoid arthritis (RA) and periodontitis. The classical interleukin-1 (IL-1) family cytokines, IL-1alpha and IL-1beta, as well as IL-18, play key roles in inflammation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service