Skip to Content
Merck
All Photos(1)

Key Documents

I7659

Sigma-Aldrich

Monoclonal Anti-Interleukin-1α antibody produced in mouse

clone 3405, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL-1α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3405, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 25 μg/mL
neutralization: suitable

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL1A(3552)

Looking for similar products? Visit Product Comparison Guide

Immunogen

purified, E. coli-derived recombinant human interleukin-1α.

Biochem/physiol Actions

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signaling through IL-1 receptor type 1. The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation. Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel Weber et al.
Science signaling, 3(105), cm1-cm1 (2010-01-21)
The interleukin-1 (IL-1) family of cytokines comprises 11 proteins (IL-1F1 to IL-1F11) encoded by 11 distinct genes in humans and mice. IL-1-type cytokines are major mediators of innate immune reactions, and blockade of the founding members IL-1alpha or IL-1beta by
H E Barksby et al.
Clinical and experimental immunology, 149(2), 217-225 (2007-06-26)
Understanding cytokine immunobiology is central to the development of rational therapies for destructive inflammatory diseases such as rheumatoid arthritis (RA) and periodontitis. The classical interleukin-1 (IL-1) family cytokines, IL-1alpha and IL-1beta, as well as IL-18, play key roles in inflammation.
Emmanuel Contassot et al.
Swiss medical weekly, 142, w13590-w13590 (2012-06-02)
Interleukin 1, one of the first cytokines discovered in the 1980s, and a potent mediator of fever, pain and inflammation, is at present experiencing a revival in biology and medicine. Whereas the mechanism of activation and secretion of interleukin 1β

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service