EP6315000015
Eppendorf® Mastercycler®
X50p, with control panel, aluminum block, size 96 wells, high pressure lid, AC/DC input 110 V / 230 V, 50-60 Hz (NA/EU)
About This Item
Recommended Products
material
aluminum block
feature
Unit can control 9 other units in network
high pressure lid
holds 1 PCR plate 8×12
unit can only be controlled
with control panel
manufacturer/tradename
Eppendorf® 6315000015
parameter
1-30 °C (gradient range)
30-99 °C (gradient temperature)
37-110 °C temp. range
4-99 °C temp. range (control range)
technique(s)
PCR: suitable
W × D × H
27.5 cm × 43 cm × 33 cm
size
96 wells
weight
11.5 kg
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Speed and enhanced PCR optimization functions, like the 2D-Gradient, make the Eppendorf Mastercycler X50 Thermocycler the ideal PCR cycler for advanced research in molecular biology. The excellent block temperature regulation gives rise to the next stage of PCR reproducibility, whereas the adaptable user management and profound documentation capabilities give peace of mind to laboratories working to specific or strict standards.
A highly intuitive touch display, low noise levels, low power consumption and the versatile flexlid concept combine to make a powerful PCR cycler. Up to 10 PCR cyclers can be combined – ideal for high throughput applications or labs with a high number of users running different assays. Should you feel you need more flexibility or throughput, up to 50 PCR cyclers can be combined in a computer-controlled network.
Product Highlights:
PCR optimization with 2D-Gradient
Denaturation temperature gradient (top to bottom by row)
Annealing temperature gradient (left to right by column)
Faster Heating and Cooling
Heating rate: up to 10°C/ sec. Eppendorf Mastercycler x50 Thermocycler
Cooling rate: 5°C/ sec.
Lid Height Adjusts Automatically
flexlid concept- the automatically adjusts to the heights of plates and tubes; allowing you to use all types of consumables
Color Touchscreen
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service