EP5495505008
Rotor for Eppendorf® 5425 Centrifuge
FA-10x5, to hold, 10 x 5.0 mL Eppendorf tubes, lid
All Photos(1)
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Recommended Products
feature
lid
to hold 10 x 5.0 mL Eppendorf tubes
FA-10x5
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495505008
parameter
15,060 max. RCF (21,300 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5425
Related Categories
General description
Rotor for Eppendorf(R) 5425 Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)