Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122194

Eppendorf® Tubes 15 mL

amber, sterile, pack of 200 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

amber tube
conical bottom
high-density polyethylene cap
polypropylene tube
screw top cap
white cap

sterility

sterile

feature

graduations
neck thread External thread

packaging

pack of 200 ea

manufacturer/tradename

Eppendorf® 0030122194

capacity

15 mL

tube diam.

17 mm

color

amber

General description

Eppendorf® Conical tubes with screw caps are universally used in a wide range of laboratory procedures and are one of the most common vessel formats. Eppendorf Conical Tubes 15 mL is an ideal solution for cell culture and cell biology applications as well as for sample preparation protocols in microbiology and molecular biology laboratories.

Application

Eppendorf® Tubes 15 mL are suitable for liquid, sample, and cell handling during centrifugation, mixing, or other applications.

Features and Benefits

  • Manufactured with premium raw material and no use of slip agents, plasticizers, and biocides to ensure the highest sample integrity, high g-Safe® centrifugation stability, and optimal sample and pellet visibility.
  • Caps with flattened and grooved sides for improved ergonomic handling and stable upright positioning, with minimum contamination risk.
  • Large writing areas with flat and light-colored caps enable reliable and safe labeling.
  • Precise dimensions support maximum compatibility with centrifuge rotors, mixers, and shakers.
  • Tested for the absence of pyrogen-, DNase-, RNase-, human and bacterial DNA.
  • Amber for UV (Ultraviolet) light-sensitive samples

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service