Skip to Content
Merck
All Photos(1)

Documents

AsPC-1

96020930, human pancreas (ascites meta), Not specified

Synonym(s):

ASPC1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AsPC-1, 96020930

biological source

human pancreas (ascites meta)

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

carcinoembryonic antigen (CEA), human pancreas associated antigen, human pancreas specific antigen, mucin.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human pancreas adenocarcinoma ascites metastasis

Cell Line Description

This cell line was derived from nude mouse xenografts initiated with cells from the ascites of a 62-year-old female Caucasian patient with cancer of the pancreas. The cells produce carcinoembryonic antigen (CEA), human pancreas associated antigen, human pancreas specific antigen and mucin.

Culture Medium

RPMI 1640 + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service