Skip to Content
Merck
All Photos(1)

Documents

FM3Ats C1.T85

87111904, mouse breast, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

FM3Ats C1.T85, 87111904

biological source

mouse breast

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H mammary carcinoma

Cell Line Description

A temperature sensitive mutant isolated from the mouse mammary cell carcinoma cell line, FM3A (Sigma Catalogue number. 87100804). Cells will not divide at 39°C (non-permissive temperature) and will grow at 33°C (permissive temperature). Electron microscope findings have shown that chromatin abnormally condenses into fragmented and compact forms in 80% of the cells at 39°C for 16 hours. This abnormal condensation does not occur at 33°C or in the FM3A cell line at either temperature.

Application

Tumourigenicity studies, chromosome condensation studies.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1-3x100,000 cells/ml; 5% CO2; 33°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service