Skip to Content
Merck
All Photos(2)

Key Documents

AV38773

Sigma-Aldrich

Anti-TSG101 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Ttumor susceptibility gene 101

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

human, horse, dog, rabbit, guinea pig, bovine, rat, goat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSG101(7251)

Immunogen

Synthetic peptide directed towards the C terminal region of human TSG101

Biochem/physiol Actions

TSG101 belongs to a group of apparently inactive homologs of ubiquitin-conjugating enzymes. TSG101 contains a coiled-coil domain that interacts with stathmin, a cytosolic phosphoprotein implicated in tumorigenesis. TSG101 may play a role in cell growth and differentiation and act as a negative growth regulator. In vitro steady-state expression of TSG101 appears to be important for maintenance of genomic stability and cell cycle regulation. Mutations and alternative splicing in TSG101 gene occur in high frequency in breast cancer. The protein encoded by this gene belongs to a group of apparently inactive homologs of ubiquitin-conjugating enzymes. The gene product contains a coiled-coil domain that interacts with stathmin, a cytosolic phosphoprotein implicated in tumorigenesis. The protein may play a role in cell growth and differentiation and act as a negative growth regulator. In vitro steady-state expression of this tumor susceptibility gene appears to be important for maintenance of genomic stability and cell cycle regulation. Mutations and alternative splicing in this gene occur in high frequency in breast cancer and suggest that defects occur during breast cancer tumorigenesis and/or progression.

Sequence

Synthetic peptide located within the following region: TIFYLGEALRRGVIDLDVFLKHVRLLSRKQFQLRALMQKARKTAGLSDLY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David E Reynolds et al.
Journal of extracellular biology, 2(6), e89-e89 (2023-05-28)
The use of mesenchymal stem cells (MSCs) in human and veterinary clinical applications has become a subject of increasing importance due to their roles in immunomodulation and regenerative processes. MSCs are especially relevant in equine medicine because they may have
Dishiwen Liu et al.
Oxidative medicine and cellular longevity, 2022, 3961495-3961495 (2022-06-10)
The exact mechanism of atrial fibrillation (AF) has been not well elucidated. Ferroptosis is an iron-dependent cell death due to excessive accumulation of peroxidized polyunsaturated fatty acids. However, the molecular mechanism underlying AF and ferroptosis has never been reported. Here
H N Gao et al.
Journal of dairy science, 102(2), 985-996 (2018-12-26)
Intestinal epithelial cells (IEC) are an important part of the intestinal barrier. Barrier function was disrupted under hypoxia, but milk-derived exosomes can regulate the intestinal barrier function. However, the mechanisms underlying the association between yak milk exosomes and hypoxia in
Angela M Gaesser et al.
Journal of the American Veterinary Medical Association, 262(S1), S97-S108 (2024-03-29)
Mesenchymal stem cell (MSC) extracellular vesicles (EVs) have emerged as a biotherapeutic for osteoarthritis; however, manufacturing large quantities is not practical using traditional monolayer (2-D) culture. We aimed to examine the effects of 3-D and 2-D culture 2 types of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service