Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503606

Sigma-Aldrich

Anti-CD9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

.MRP-1, CD9 antigen, Leukocyte antigen MIC3, Motility-related protein, p24

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 25 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD9(928)

General description

Anti-CD9 Antibody detects endogenous levels of total CD9 protein.
CD9 (CD9 molecule) codes for a transmembrane protein,tetraspanin. It is a member of tetraspanin family. It is mapped to human chromosome 12p13.31.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD9.

Immunogen Range: 101-150

Biochem/physiol Actions

Tetraspanin coded by CD9 (CD9 molecule) participates in tumor cell invasion, apoptosis and resistance to chemotherapy. It also participates in EGFR (epidermal growth factor receptor) and FGFR (fibroblast growth factor receptor) signaling pathways.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparing cDNA and oligonucleotide array data: concordance of gene expression across platforms for the NCI-60 cancer cells
Lee JK, et al.
Genome Biology (2003)
Yuxuan Fu et al.
MedComm, 4(5), e384-e384 (2023-09-27)
Extracellular vesicles (EVs), including exosomes and microvesicles (MVs), are released by most cell types into the extracellular space and represent the pathophysiological condition of their source cells. Recent studies demonstrate that EVs derived from infected cells and tumors contribute to
Yuxuan Fu et al.
Journal of controlled release : official journal of the Controlled Release Society, 335, 584-595 (2021-06-06)
The worldwide spread of COVID-19 highlights the urgent need for an efficient approach to rapidly develop therapeutics and prophylactics against SARS-CoV-2. Extracellular vesicle(EVs) are recognized and endocytosed by tissue cells via specific interactions between surface membrane proteins, where after they
Yuxuan Fu et al.
PLoS pathogens, 19(1), e1011090-e1011090 (2023-01-13)
Specific virus-receptor interactions are important determinants in viral host range, tropism and pathogenesis, influencing the location and initiation of primary infection as well as viral spread to other target organs/tissues in the postviremic phase. Coxsackieviruses of Group B (CVB) and
Zeyan Liang et al.
Neural regeneration research, 19(10), 2259-2269 (2024-03-15)
JOURNAL/nrgr/04.03/01300535-202410000-00027/figure1/v/2024-02-06T055622Z/r/image-tiff Spinal cord injury is a disabling condition with limited treatment options. Multiple studies have provided evidence suggesting that small extracellular vesicles (SEVs) secreted by bone marrow mesenchymal stem cells (MSCs) help mediate the beneficial effects conferred by MSC transplantation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service