Skip to Content
Merck
All Photos(1)

Key Documents

H3410

Sigma-Aldrich

Hydrogen peroxide solution

contains inhibitor, 30 wt. % in H2O, meets USP testing specifications

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

vapor pressure

23.3 mmHg ( 30 °C)

Assay

≤32%

form

liquid

reaction suitability

reagent type: oxidant

concentration

30 wt. % in H2O

density

1.11 g/mL at 25 °C

cation traces

heavy metals: <5 ppm

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 30% solution of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and by-product is water. This product is a high quality pharmacopoeia product that meets the testing specifications of USP (United States Pharmacopoeia). It can be employed in research and pilot studies.

Application

Hydrogen peroxide solution has been used:
  • To prepare a sterilant solution along with peracetic acid that is employed in a novel wash process for bone allograft.
  • In combination with 3,3′,5,5′-tetramethylbenzidine dihydrochloride (TMB) to prepare horseradish peroxidase (HRP)-conjugated gold nanoparticles (GNPs).
  • For the immunostaining of NeuN (a soluble nuclear protein) expressing neurons.

Preparation Note

Stabilized

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinghua Sun et al.
Annals of biomedical engineering, 40(10), 2131-2139 (2012-04-26)
Gold-gold sulfide nanoparticles (GGS-NPs) fabricated from chloroauric acid and sodium thiosulfate show unique near infrared (NIR) absorption that renders them as a promising candidate for photothermal cancer therapy. To improve targeting efficiency, we developed a versatile method to allow ordered
Strukul G.
Catalytic Oxidations with Hydrogen Peroxide as Oxidant., 5-7 null
C A Smith et al.
Journal of tissue engineering and regenerative medicine, 9(5), 595-604 (2014-06-20)
Fresh-frozen biological allograft remains the most effective substitute for the 'gold standard' autograft, sharing many of its osteogenic properties but, conversely, lacking viable osteogenic cells. Tissue engineering offers the opportunity to improve the osseointegration of this material through the addition
Petra Sántha et al.
Frontiers in molecular neuroscience, 8, 88-88 (2016-02-03)
Stress is well-known to contribute to the development of both neurological and psychiatric diseases. While the role of the blood-brain barrier is increasingly recognized in the development of neurodegenerative disorders, such as Alzheimer's disease, dysfunction of the blood-brain barrier has
Koen van der Maaden et al.
Pharmaceutical research, 31(7), 1846-1854 (2014-01-29)
The aim of the study was to develop a cheap and fast method to produce hollow microneedles and an applicator for injecting vaccines into the skin at a pre-defined depth and test the applicability of the system for dermal polio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service