Skip to Content
Merck
All Photos(4)

Documents

239283

Sigma-Aldrich

Potassium hydrogenfluoride

99%

Synonym(s):

Potassium bifluoride, Potassium hydrogen difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHF2
CAS Number:
Molecular Weight:
78.10
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

99%

form

lumps
powder or crystals

mp

239 °C (lit.)

density

2.37 g/mL at 25 °C (lit.)

SMILES string

[F-].[K+].F[H]

InChI

1S/2FH.K/h2*1H;/q;;+1/p-1

InChI key

VBKNTGMWIPUCRF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis of 4′-deoxy-4′-fluorokanamycin A and B: This study discusses the synthetic applications of Potassium hydrogenfluoride in the development of fluorinated antibiotics, highlighting its role in selective fluorination processes (Takahashi Y et al., 1992).
  • Reaction of 2-deoxy-6-O-[2,3-dideoxy-4,6-O-isopropylidene-2,3-(N-tosylepimino)-alpha-D-mannopyranosyl]-4,5-O-isopropylidene-1,3-di-N-tosylstreptamine with potassium hydrogenfluoride: This article explores the chemical behavior of Potassium hydrogenfluoride under specific synthetic conditions, providing insights into its role in complex organic transformations (Kobayashi Y et al., 1992).
  • A synthetic study of methyl 3-deoxy-3-fluoro-alpha-D-glucopyranosides from methyl 2,3-anhydro-alpha-D-allopyranosides, and synthesis of 3′-deoxy-3′-fluorokanamycin A and 3′-chloro-3′-deoxykanamycin A: This research delves into the synthesis of modified sugars using Potassium hydrogenfluoride, illustrating its utility in the creation of novel glycoside structures, which are important in drug development (Umemura E et al., 1992).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium Hydrogen Fluoride.
Molander GA, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2012)
Reaction of 2-deoxy-6-O-[2,3-dideoxy-4,6-O-isopropylidene-2,3- (N-tosylepimino)-alpha-D-mannopyranosyl]-4,5-O-isopropylidene-1,3-di-N- tosylstreptamine with potassium hydrogenfluoride.
Y Kobayashi et al.
Carbohydrate research, 229(2), 363-368 (1992-05-22)
Gary A Molander et al.
The Journal of organic chemistry, 73(17), 6841-6844 (2008-08-07)
A number of alkynyl pinacolboronates bearing various functional groups were synthesized according to literature methods. These were then stereoselectively reduced to the cis-alkenyl pinacolboronates via hydroboration with dicyclohexylborane followed by chemoselective protodeboronation using acetic acid. Treatment with potassium hydrogen fluoride
Zhibo Liu et al.
Nature protocols, 10(9), 1423-1432 (2015-08-28)
Herein we present a general protocol for the functionalization of biomolecules with an organotrifluoroborate moiety so that they can be radiolabeled with aqueous (18)F fluoride ((18)F(-)) and used for positron emission tomography (PET) imaging. Among the β(+)-emitting radionuclides, fluorine-18 ((18)F)
Matthew J Hesse et al.
Angewandte Chemie (International ed. in English), 51(50), 12444-12448 (2012-11-07)
E-allylic boronic esters undergo a highly diastereoselective protodeboronation with TBAF⋅3 H(2)O to give Z-trisubstituted alkenes. The selectivity can be switched to give predominantly the E-alkene instead by using KHF(2)/TsOH (see scheme). The utility of the methodology has been illustrated in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service