Merck
  • Diastereodivergent synthesis of trisubstituted alkenes through protodeboronation of allylic boronic esters: application to the synthesis of the Californian red scale beetle pheromone.
Diastereodivergent synthesis of trisubstituted alkenes through protodeboronation of allylic boronic esters: application to the synthesis of the Californian red scale beetle pheromone.
Angewandte Chemie (International ed. in English) (2012-11-07)
Matthew J Hesse, Craig P Butts, Christine L Willis, Varinder K Aggarwal
ABSTRACT
E-allylic boronic esters undergo a highly diastereoselective protodeboronation with TBAF⋅3 H(2)O to give Z-trisubstituted alkenes. The selectivity can be switched to give predominantly the E-alkene instead by using KHF(2)/TsOH (see scheme). The utility of the methodology has been illustrated in a short synthesis of a component of the sex pheromone of the Californian red scale beetle.
Sigma-Aldrich
Potassium hydrogenfluoride, 99%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Potassium hydrogenfluoride solution, 3.0 M in H2O