Skip to Content
Merck
All Photos(1)

Key Documents

P1700000

Pilocarpine nitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pilocarpine nitrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HNO3
CAS Number:
Molecular Weight:
271.27
Beilstein:
4171406
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pilocarpine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[N+]([O-])=O.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.HNO3/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;2-1(3)4/h5,7-8,10H,3-4,6H2,1-2H3;(H,2,3,4)/t8-,10-;/m0./s1

InChI key

PRZXEPJJHQYOGF-GNAZCLTHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pilocarpine nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barina Aqil et al.
Archives of pathology & laboratory medicine, 138(7), 920-922 (2014-02-27)
All positive screening of newborns for cystic fibrosis using the dried blood spot 2-tiered immunoreactive trypsinogen/DNA method requires subsequent sweat chloride testing for confirmation. Obtaining an adequate volume of sweat to measure chloride is a challenge for many cystic fibrosis
Marcele Jardim Pimentel et al.
Anticancer research, 34(4), 1993-1999 (2014-04-03)
There is controversy concerning the effect of pilocarpine in the reversal of radio-induced xerostomia; however, the tests are usually performed at the end of radiotherapy. The present study evaluated the radioprotective effects of pilocarpine when ingested during radiation treatment. Eleven
G Asuntha et al.
Indian journal of experimental biology, 53(1), 31-35 (2015-02-14)
Argemone mexicana (L.) has a role in the treatment of epileptic disorders in Indian traditional system of medicine. We studied its effect on induced status epilepticus (SE) and oxidative stress in rats. SE was induced in male albino rats by
Lin He et al.
Experimental eye research, 125, 244-255 (2014-07-02)
This study was undertaken to understand the role of anterior chamber pressure (ACP) during pharmacological and Edinger-Westphal (EW) stimulated accommodation in anesthetized monkeys. Experiments were performed on one iridectomized eye each of 7 anesthetized adolescent rhesus monkeys. Accommodation was induced
Yuan-yuan Zhang et al.
Acta pharmacologica Sinica, 35(9), 1137-1149 (2014-08-05)
The non-neuronal acetylcholine system (NNAS) in endothelial cells participates in modulating endothelial function, vascular tone, angiogenesis and inflammation, thus plays a critical role in cardiovascular diseases. In this study, we used a proteomic approach to study potential downstream receptor-effectors of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service