Merck
  • Implementation of a quality improvement program to improve sweat test performance in a pediatric hospital.
Implementation of a quality improvement program to improve sweat test performance in a pediatric hospital.
Archives of pathology & laboratory medicine (2014-02-27)
Barina Aqil, Aaron West, Michael Dowlin, Estella Tam, Cristy Nordstrom, Gregory Buffone, Sridevi Devaraj
ABSTRACT
All positive screening of newborns for cystic fibrosis using the dried blood spot 2-tiered immunoreactive trypsinogen/DNA method requires subsequent sweat chloride testing for confirmation. Obtaining an adequate volume of sweat to measure chloride is a challenge for many cystic fibrosis centers across the nation. The standard for patients older than 3 months is less than 5% quantity not sufficient (QNS) and for patients 3 months or younger is less than 10% QNS. To set up a quality improvement (QI) program for sweat testing to improve QNS rates using the Wescor Macroduct (Wescor, Inc, Logan, Utah) method at Texas Children's Hospital's laboratory, Houston, Texas. Single-center study. Quantity not sufficient rates were evaluated for 4 months before and 8 months after implementation of the QI program for patients aged 3 months or younger and those older than 3 months. The QI program included changes in technician training, service, site of collection, mode of collection, weekly review, and forms to screen patients for medications that may alter sweat production. A marked improvement was observed in the rates of QNS, which declined considerably from 16.7% to 8.5% (≤3 months old) and from 9.3% to 2.2% (>3 months old) after implementation of the QI initiative in both age categories. This report demonstrates the effectiveness of the QI program in significantly improving QNS rates in sweat chloride testing in a pediatric hospital.
Product Description
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Pilocarpine nitrate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications
Pilocarpine nitrate, European Pharmacopoeia (EP) Reference Standard