Skip to Content
Merck
All Photos(1)

Key Documents

D0940000

Diazepam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Chloro-1-methyl-5-phenyl-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-2807

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diazepam

manufacturer/tradename

EDQM

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(=O)CN=C(c2ccccc2)c3cc(Cl)ccc13

InChI

1S/C16H13ClN2O/c1-19-14-8-7-12(17)9-13(14)16(18-10-15(19)20)11-5-3-2-4-6-11/h2-9H,10H2,1H3

InChI key

AAOVKJBEBIDNHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diazepam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Diazepam is a benzodiazepine anxiolytic agent. It serves as a prototype ligand for the GABAA receptor benzodiazepine modulatory site.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ryota Inokuchi et al.
Medicine, 94(7), e555-e555 (2015-02-24)
Administering diazepam intravenously or rectally in an adult with status epilepticus can be difficult and time consuming. The aim of this study was to examine whether intranasal diazepam is an effective alternative to intravenous diazepam when treating status epilepticus. We
Jean-Baptiste Faure et al.
Epilepsia, 55(5), 644-653 (2014-03-14)
Temporal lobe epilepsy is a relatively frequent, invalidating, and often refractory neurologic disorder. It is associated with cognitive impairments that affect memory and executive functions. In the rat lithium-pilocarpine temporal lobe epilepsy model, memory impairment and anxiety disorder are classically
Matteo Cerri et al.
PloS one, 9(11), e112849-e112849 (2014-11-15)
Neurons within the lateral hypothalamus (LH) are thought to be able to evoke behavioural responses that are coordinated with an adequate level of autonomic activity. Recently, the acute pharmacological inhibition of LH has been shown to depress wakefulness and promote
M Iijima et al.
British journal of pharmacology, 171(14), 3511-3525 (2014-03-25)
Vasopressin V1B receptor antagonists may be effective for the treatment of depression and anxiety and the objective of this study was to characterize the pharmacological profiles of two newly synthesized arginine vasopressin receptor 1B (V1B receptor) antagonists, TASP0233278 and TASP0390325.
A N J Pietersen et al.
Journal of neurophysiology, 112(6), 1421-1438 (2014-06-13)
Visual perception requires integrating signals arriving at different times from parallel visual streams. For example, signals carried on the phasic-magnocellular (MC) pathway reach the cerebral cortex pathways some tens of milliseconds before signals traveling on the tonic-parvocellular (PC) pathway. Visual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service